missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(hydroxymethyl)propane, TRC

Product Code. 30135870
Change view
Click to view available options
Quantity:
50 g
500 g
Unit Size:
500g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135870 500 g 500g
17962645 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135870 Supplier Toronto Research Chemicals Supplier No. TRCT876340500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-99-6
Formula Weight 134.0943
InChI Formula InChI=1S/C6H14O3/c1-2-6(3-7,4-8)5-9/h7-9H,2-5H2,1H3
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula C6 H14 O3
Molecular Weight (g/mol) 134.17
SMILES CCC(CO)(CO)CO
Synonym 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 1,1,1-Tri(hydroxymethyl)propane, 1,1,1-Trimethylolpropane, 1,1,1-Tris(hydroxymethyl)propane, 2,2-Bis(hydroxymethyl)-1-butanol, 2-(Hydroxymethyl)-2-ethyl-1,3-propanediol, 2-Ethyl-2-(hydroxymethyl)propanediol, 2-Ethyl-2-hydroxymethyl-1,3-propandiol, Addolink TR, Ethriol, Ethyltrimethylolmethane, Ettriol, NSC 3576, Propane, 1,1,1-tris(hydroxymethyl)-, RC Crosslinker TR, T 0480, TMP, TMP (alcohol), Trimethylolpropane, Tris(hydroxymethyl)propane
Chemical Name or Material 1,1,1-Tri(hydroxymethyl)propane

Specifications

Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.