accessibility menu, dialog, popup

UserName

CAS RN 77-99-6

CH3HOHOHO

CAS RN 77-99-6

IUPAC Name: 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Synonyms: trimethylolpropane
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: ZJCCRDAZUWHFQH-UHFFFAOYSA-N
SMILES: CCC(CO)(CO)CO
Boiling Point 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)