missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(hydroxymethyl)propane, TRC

Product Code. 17962645
Change view
Click to view available options
Quantity:
50 g
500 g
Unit Size:
500g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962645 50 g 50g
30135870 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17962645 Supplier Toronto Research Chemicals Supplier No. TRCT87634050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-99-6
Formula Weight 134.0943
InChI Formula InChI=1S/C6H14O3/c1-2-6(3-7,4-8)5-9/h7-9H,2-5H2,1H3
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.17
SMILES OCC(CO)(CO)CC
Synonym 1,1,1-Tri(hydroxymethyl)propane, 1,1,1-Trimethylolpropane, 2,2-Bis(hydroxymethyl)-1-butanol, 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 2-Ethyl-2-(hydroxymethyl)propanediol, Addolink TR, Ethriol, Ethyltrimethylolmethane, Ettriol, NSC 3576, RC Crosslinker TR, TMP, TMP (alcohol), Trimethylolpropane, Tris(hydroxymethyl)propane, 2-Ethyl-2-(hydroxymethyl)propane-1,3-diol
Chemical Name or Material 1,1,1-Tris(hydroxymethyl)propane

Specifications

Quantity 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.