Learn More
TSH beta Antibody (154), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62385AF594
This item is not returnable.
View return policy
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| TSH beta | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| 154 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Native Human beta-TSH | |
| 0.5 mL | |
| Cancer, Signal Transduction | |
| 7252 | |
| Store at 4C in the dark. | |
| IgG1 |