Learn More
Trajan Scientific and Medical™ 0.36mm PEEKsil™ Tubing
Click to view available options
Overall Length (Metric):
100 m
150 m
250 m
50 m
500 m
Diameter (Metric) Inner:
0.025 mm
0.05 mm
Unit Size:
Pack of 2
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
| Type | Tubing |
| Includes | 2/Pk. |
| Diameter (Metric) Inner | 0.025 mm |
| Overall Length (Metric) | 50 m |