missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Antibody (J2) [Janelia Fluor™ 635], Novus Biologicals™
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Dot Blot,Affinity Purification,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | dsRNA |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Dot Blot, Affinity Purification, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | J2 |
| Conjugate | Janelia Fluor 635 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Double-stranded RNA |
| Host Species | Mouse |
| Immunogen | Female DBA/2 mice were injected intraperitonially with a mixture of 50 μg L-dsRNA and 75 μg methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?