missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 Antibody (MC-480), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Marke: Novus Biologicals NB100-1831
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse, Rat, Chimpanzee samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spezifikation
| SSEA-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Simple Western 1:10, Flow Cytometry 2.5 ug/million cells, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200 | |
| alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
| Mouse | |
| IgM purified | |
| RUO | |
| Primary | |
| Human and mouse. This antibody recognizes SSEA1 in mouse EC and ES cells but not human, but SSEA1 is expressed by some human differentiated cell types (ie: trophoblasts). Reactivity with Rat is reported by Kim et al. 2007 (PMID 17548530). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| MC-480 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FUT4 | |
| F9 tetracarcinoma stem cells (X-irradiated). | |
| 0.1 mL | |
| Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| 2526 | |
| Human, Mouse, Rat, Chimpanzee | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur