missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (1168B), PE/Atto594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8634PEATT594
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Monoclonal antibody specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Pref-1/DLK1/FA1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1168B | |
| Flow Cytometry | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Mouse myeloma cell line NS0-derived recombinant mouse Pref-1/DLK1/FA1, Ala24-Gln305, Accession # Q09163 | |
| 0.1 mL | |
| Signal Transduction | |
| 8788 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction