missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NOB1 Monoclonal antibody specifically detects NOB1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NOB1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1C12 |
| Conjugate | FITC |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1 |
| Host Species | Mouse |
| Immunogen | Human recombinant protein fragment corresponding to amino acids 2-234 of human NOB1(NP_054781) produced in E.coli. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?