missing translation for 'onlineSavingsMsg'
Learn More

Muscarinic Acetylcholine Receptor M1/CHRM1 Antibody, Novus Biologicals™

Product Code. 18749953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749953 0.1 mL 0.1mL
18451292 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18749953 Supplier Novus Biologicals Supplier No. NBP187466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

Muscarinic Acetylcholine Receptor M1/CHRM1 Polyclonal specifically detects Muscarinic Acetylcholine Receptor M1/CHRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Dilution Western Blot Reported in scientific literature (PMID: 24658304)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence Validated from a verified customer review., Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen Reported in scientific literature (PMID: 24658304)., Immunohistochemistry Free-Floating Reported in scientific literature (PMID: 31785263).
Gene Alias cholinergic receptor, muscarinic 1, HM1, M1, M1R, MGC30125, muscarinic acetylcholine receptor M1
Quantity 0.1 mL
Regulatory Status RUO
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.