missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Musashi-2 Antibody (960121) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3255I
This item is not returnable.
View return policy
Description
Musashi-2 Monoclonal antibody specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Musashi-2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 960121 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| FLJ36569, MGC3245, MSI2H, musashi homolog 2 (Drosophila), musashi-2, RNA-binding protein Musashi homolog 2 | |
| E. coli-derived recombinant human Musashi-2, Met1-His328, Accession # Q96DH6 | |
| 0.1 mL | |
| Neuronal Cell Markers | |
| 124540 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction