missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX2 Antibody (CL3202) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42398JF646
This item is not returnable.
View return policy
Description
SSX2 Monoclonal antibody specifically detects SSX2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| SSX2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Cancer/testis antigen 5.2, cancer/testis antigen family 5, member 2a, CT5.2a, HD21, HOM-MEL-40, MGC119055, MGC15364, MGC3884, protein SSX2, sarcoma, synovial, X-chromosome-related 2, SSX, SSX2A, SSX2B, synovial sarcoma, X breakpoint 2B, synovial sarcoma, X breakpoint 2CT5.2, Tumor antigen HOM-MEL-40 | |
| This antibody was developed using a recombinant protein derived from Q16385, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 6757 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL3202 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction