missing translation for 'onlineSavingsMsg'
Learn More

HMG-CoA Reductase/HMGCR Antibody (CL0260) [Alexa Fluor™ 405], Novus Biologicals™

Product Code. 30293118 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30293118 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30293118 Supplier Novus Biologicals Supplier No. NBP261617AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMG-CoA Reductase/HMGCR Monoclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMG-CoA Reductase/HMGCR
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone CL0260
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias 3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P04035, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3156
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.