Learn More
LL-37 Antibody (B-F57), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18122MFV500
This item is not returnable.
View return policy
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Western Blot, ELISA | |
| RUO | |
| Innate Immunity | |
| Purified |