missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JPH4 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41095R
This item is not returnable.
View return policy
Description
JPH4 Polyclonal antibody specifically detects JPH4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| JPH4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| DyLight 550 | |
| JP4, JP-4, JPHL1, Junctophilin 4, Junctophilin Like 1, junctophilin-4, Junctophilin-Like 1 Protein, KIAA1831 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human JPH4. The immunogen is located within amino acids 410 - 460 of JPH4. Amino Acid Squence: LEAPFRRPRQDSEGSDT | |
| 0.1 mL | |
| Cell Biology, Neuroscience | |
| 84502 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction