missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RA/IL-17R Antibody (133617), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB177H
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF, Gene Knock-Out
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Neutralization, CyTOF, Gene Knock-Out | |
| 133617 | |
| Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, Knockout Validated | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RA/IL-17 R, Leu33-Trp320, Accession # Q96F46 | |
| 0.1 mL | |
| Innate Immunity | |
| 23765 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction