missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Antibody (OTI1A4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71198CL1
This item is not returnable.
View return policy
Description
FOXO4 Monoclonal antibody specifically detects FOXO4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| FOXO4 | |
| Monoclonal | |
| CoraFluor 1 | |
| AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7 | |
| Human recombinant protein fragment corresponding to amino acids 206-505 of human FOXO4 (NP_005929) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 4303 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1A4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction