Learn More
Cannabinoid R1/CB1/CNR1 Antibody (368302), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3834PEATT594
Product Code. 30039667
  • 730.00€ / 0.10mL
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cannabinoid R1/CB1/CNR1
Monoclonal
PE/ATTO 594
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
368302
Flow Cytometry
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167
0.1 mL
Cancer, GPCR, Neuroscience, Neurotransmission, Vision
1268
Store at 4C in the dark. Do not freeze.
IgG2a