missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Chorionic Gonadotropin beta Chain (hCG beta) His Protein

Product Code. 18641779 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18641779

Brand: Novus Biologicals™ NBP2595450.05mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 21-165 of Human Chorionic Gonadotropin beta Chain (HCG beta) The Recombinant Human Chorionic Gonadotropin beta Chain (hCG beta) Protein is derived from E. coli. The Recombinant Human Chorionic Gonadotropin beta Chain (hCG beta) Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25mg/mL
For Use With (Application) PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 1082
Molecular Weight (g/mol) 16.6 kDa
Name Recombinant Human Chorionic Gonadotropin beta Chain (HCG beta) Protein
Purification Method >90%, by SDS-PAGE
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
Gene Alias CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hC
Gene Symbol CGB
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Species Human
Recombinant Recombinant
Form Purified
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.