Learn More
0.01 M Phosphoric Acid and 0.01 M Monobasic Sodium Phosphate Buffered Eluent, pH 2.5, In Accordance with United States Pharmacopoeia (USP), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
| Standard Concentration | 0.01 M |
| Item Description | pH 2.5 buffered eluent |
| Quantity | 1 L |
| Form | Liquid |
| For Use With | Mobile Phase Eluent |