accessibility menu, dialog, popup

UserName

CAS RN 900572-37-4

NHON

CAS RN 900572-37-4

IUPAC Name: 2-(piperidin-4-yloxy)benzonitrile
Synonyms: 2-(piperidin-4-yloxy)benzonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: OIFRRCRLKBGBKR-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1OC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results