accessibility menu, dialog, popup

UserName

CAS RN 885266-74-0

HNFBr

CAS RN 885266-74-0

IUPAC Name: 2-(4-bromophenyl)-5-fluoro-1H-indole
Synonyms: 2-(4-bromophenyl)-5-fluoro-1H-indole
Molecular Weight (g/mol): 290.14
Molecular Formula: C14H9BrFN
InChi Key: XAKLXLIHCVKNII-UHFFFAOYSA-N
SMILES: FC1=CC=C2NC(=CC2=C1)C1=CC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results