CAS RN 859-18-7
CAS RN 859-18-7
IUPAC Name:
hydrogen (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide chloride
Synonyms:
hydrogen lincomycin chloride
Molecular Weight (g/mol):
443
Molecular Formula:
C18H35ClN2O6S
InChi Key:
POUMFISTNHIPTI-BOMBIWCESA-N
SMILES:
[H+].[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
1
–
3
of
3
results
1
Lincomycin hydrochloride
CAS: 859-18-7 Molecular Formula: C18H35ClN2O6S Molecular Weight (g/mol): 442.996 MDL Number: MFCD00058237 InChI Key: POUMFISTNHIPTI-OXVORJCQSA-N PubChem CID: 124080936 IUPAC Name: (2R,4R)-N-[(1R,2R)-2-hydroxy-1-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride SMILES: CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)O.Cl
2
Lincomycin, hydrochloride, ≥82%, MP Biomedicals™
Lincomycin, Hydrochloride
3
Lincomycin Hydrochloride Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.