CAS RN 847749-37-5
CAS RN 847749-37-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pyrisoxazole, TRC
CAS: 847749-37-5 Molecular Formula: C16 H17 Cl N2 O Molecular Weight (g/mol): 288.772 Synonym: 3-[5-(4-Chlorophenyl)-2,3-dimethyl-3-isoxazolidinyl]pyridine,3-[5-(4-Chlorophenyl)-2,3-dimethyl-1,2-oxazolidin-3-yl]pyridine,3-[5-(4-Chlorophenyl)-2,3-dimethylisooxazolidin-3-yl]pyridine,3-[5-(4-Chlorophenyl)-2,3-dimethylisoxazolidin-3-yl]pyridine,Dingjunezuo,Pyrisoxazole,SYP-Z 048 IUPAC Name: 5-(4-chlorophenyl)-2,3-dimethyl-3-pyridin-3-yl-1,2-oxazolidine SMILES: CN1OC(CC1(C)c2cccnc2)c3ccc(Cl)cc3