missing translation for 'onlineSavingsMsg'
Learn More

Pyrisoxazole, TRC

Product Code. 17972245
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972245 10 mg 10mg
17963595 25 mg 25mg
17963605 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972245 Supplier Toronto Research Chemicals Supplier No. TRCP99880510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 847749-37-5
Molecular Formula C16 H17 Cl N2 O
Molecular Weight (g/mol) 288.772
Synonym 3-[5-(4-Chlorophenyl)-2,3-dimethyl-3-isoxazolidinyl]pyridine, 3-[5-(4-Chlorophenyl)-2,3-dimethyl-1,2-oxazolidin-3-yl]pyridine, 3-[5-(4-Chlorophenyl)-2,3-dimethylisooxazolidin-3-yl]pyridine, 3-[5-(4-Chlorophenyl)-2,3-dimethylisoxazolidin-3-yl]pyridine, Dingjunezuo, Pyrisoxazole, SYP-Z 048
IUPAC Name 5-(4-chlorophenyl)-2,3-dimethyl-3-pyridin-3-yl-1,2-oxazolidine
SMILES CN1OC(CC1(C)c2cccnc2)c3ccc(Cl)cc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H17ClN2O/c1-16(13-4-3-9-18-11-13)10-15(20-19(16)2)12-5-7-14(17)8-6-12/h3-9,11,15H,10H2,1-2H3
Formula Weight 288.1029
Percent Purity >95%
Chemical Name or Material Pyrisoxazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.