accessibility menu, dialog, popup

UserName

CAS RN 84199-98-4

H3COON

CAS RN 84199-98-4

IUPAC Name: methyl 3-(pyridin-3-yl)propanoate
Synonyms: methyl 3-(pyridin-3-yl)propanoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: FQRRQPZYIYAPAV-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CN=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results