accessibility menu, dialog, popup

UserName

OClFFF

CAS RN 83279-38-3

IUPAC Name: 3-chloro-4-(trifluoromethyl)benzaldehyde
Synonyms: 3-chloro-4-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: UTSBZTCYCRVHCK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results