accessibility menu, dialog, popup

UserName

HOO

CAS RN 828-51-3

IUPAC Name: adamantane-1-carboxylic acid
Synonyms: adamantanecarboxylic acid
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: JIMXXGFJRDUSRO-UHFFFAOYSA-N
SMILES: OC(=O)C12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)