accessibility menu, dialog, popup

UserName

CAS RN 82560-54-1

H3COONSNCH3OOCH3H3COCH3H3C

CAS RN 82560-54-1

IUPAC Name: ethyl 3-{[({[(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)oxy]carbonyl}(methyl)amino)sulfanyl](propan-2-yl)amino}propanoate
Synonyms: oncol benfuracarb (iso) benfuracarb oncol 5G furacon epa pesticide chemical code 123201 caswell no. 349C aminofuracarb
Molecular Weight (g/mol): 410.53
Molecular Formula: C20H30N2O5S
InChi Key: FYZBOYWSHKHDMT-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN(SN(C)C(=O)OC1=CC=CC2=C1OC(C)(C)C2)C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results