accessibility menu, dialog, popup

UserName

CAS RN 824-54-4

CH3OBr

CAS RN 824-54-4

IUPAC Name: 2-bromo-4-methylbenzaldehyde
Synonyms: 2-bromo-4-methylbenzaldehyde
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: MUZMDYCVUCMIDC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results