missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylbenzaldehyde, 95%

Product Code. 15443717
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15443717

Brand: Thermo Scientific Alfa Aesar H27558.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-54-4 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 824-54-4
Melting Point 33°C to 37°C
Flash Point >110°C (230°F)
Molecular Formula C8H7BrO
MDL Number MFCD04039890
Quantity 1 g
Sensitivity Air Sensitive
Synonym benzaldehyde, 2-bromo-4-methyl, acmc-209pp2, ksc497q8b, tpc-i119, 2-2-methoxyphenyl-2-methylamino cyclohexanone, 2-bromo-4-methylbenzaldehyde, technical grade
InChI Key MUZMDYCVUCMIDC-UHFFFAOYSA-N
SMILES CC1=CC=C(C=O)C(Br)=C1
IUPAC Name 2-bromo-4-methylbenzaldehyde
Molecular Weight (g/mol) 199.05
PubChem CID 4645292
Formula Weight 199.05
Percent Purity 95%
Chemical Name or Material 2-Bromo-4-methylbenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.