CAS RN 82034-46-6
CAS RN 82034-46-6
IUPAC Name:
chloromethyl 1-[(ethoxycarbonyl)oxy]-10-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1-carboxylate
Synonyms:
loteprednol
Molecular Weight (g/mol):
466.96
Molecular Formula:
C24H31ClO7
InChi Key:
DMKSVUSAATWOCU-UHFFFAOYNA-N
SMILES:
CCOC(=O)OC1(CCC2C3CCC4=CC(=O)C=CC4(C)C3C(O)CC12C)C(=O)OCCl
1
–
2
of
2
results
1
Loteprednol Etabonate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Thermo Scientific Chemicals Loteprednol etabonate
CAS: 82034-46-6 Molecular Formula: C24H31ClO7