accessibility menu, dialog, popup

UserName

CAS RN 81405-85-8

CH3OOH3CNH3CH3CCH3ONH

CAS RN 81405-85-8

IUPAC Name: methyl 4-methyl-2-[4-methyl-5-oxo-4-(propan-2-yl)-4,5-dihydro-1H-imidazol-2-yl]benzoate
Synonyms: methyl 2-(4-isopropyl-4-methyl-5-oxo-1H-imidazol-2-yl)-4-methylbenzoate
Molecular Weight (g/mol): 288.35
Molecular Formula: C16H20N2O3
InChi Key: FFCCBBNQPIMUJI-UHFFFAOYNA-N
SMILES: COC(=O)C1=CC=C(C)C=C1C1=NC(C)(C(C)C)C(=O)N1
Concentration 
  • (1)
Industry Type 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results