accessibility menu, dialog, popup

UserName

CAS RN 814-29-9

H3CPOH3CH3C

CAS RN 814-29-9

IUPAC Name: 1-(dibutylphosphoryl)butane
Synonyms: tributylphosphine oxide
Molecular Weight (g/mol): 218.32
Molecular Formula: C12H27OP
InChi Key: MNZAKDODWSQONA-UHFFFAOYSA-N
SMILES: CCCCP(=O)(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results