CAS RN 80474-45-9
CAS RN 80474-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-3-oxo-17-(propanoyloxy)androsta-1,4-diene-17beta-carbothioic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.