accessibility menu, dialog, popup

UserName

CAS RN 80-75-1

H3CO(S)(S)O(R)H3C(S)(R)OH(S)H3C

CAS RN 80-75-1

IUPAC Name: (3aS,3bS,9aR,9bS,10R,11aS)-1-acetyl-10-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: duralutin 11a-hydroxyprogesterone pergestron prodrox pro-depo pregn-4-en-11a-ol-3,20-dione hy-gestrone 11a-hydroxy-progesterone hyprogest 4-pregnene-11a-ol-3,20-dione
Molecular Weight (g/mol): 330.47
Molecular Formula: C21H30O3
InChi Key: BFZHCUBIASXHPK-ODYOLWGQSA-N
SMILES: CC(=O)C1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@H](O)C[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results