accessibility menu, dialog, popup

UserName

CAS RN 80-63-7

CH3OOClH2C

CAS RN 80-63-7

IUPAC Name: methyl 2-chloroprop-2-enoate
Synonyms: methyl 2-chloroacrylate
Molecular Weight (g/mol): 120.53
Molecular Formula: C4H5ClO2
InChi Key: AWJZTPWDQYFQPQ-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results