accessibility menu, dialog, popup

UserName

CAS RN 79560-19-3

OClCl

CAS RN 79560-19-3

IUPAC Name: 4-(3,4-dichlorophenyl)-1,2,3,4-tetrahydronaphthalen-1-one
Synonyms: 4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol): 291.17
Molecular Formula: C16H12Cl2O
InChi Key: JGMBHJNMQVKDMW-UHFFFAOYNA-N
SMILES: ClC1=C(Cl)C=C(C=C1)C1CCC(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results