accessibility menu, dialog, popup

UserName

CAS RN 789-02-6

ClClClClCl

CAS RN 789-02-6

IUPAC Name: 1-chloro-4-[2,2,2-trichloro-1-(2-chlorophenyl)ethyl]benzene
Synonyms: o,p'-DDT (R)-isomer o,p'-DDT ortho,para'-DDT (R)-o,p'-DDT o,p'-DDT, (R)- o,p-DDT (r)-isomer o,p'-ddt o,p'-DDT, (R)-isomer O,P'-ddt o,p'-ddt, (r)-isomer
Molecular Weight (g/mol): 354.48
Molecular Formula: C14H9Cl5
InChi Key: CVUGPAFCQJIYDT-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C(C1=CC=CC=C1Cl)C(Cl)(Cl)Cl
Solvent or Matrix 
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (6)
  • (1)
  • (4)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)