CAS RN 7790-58-1
CAS RN 7790-58-1
IUPAC Name:
dipotassium tellurite
Synonyms:
dipotassium tellurite
Molecular Weight (g/mol):
253.79
Molecular Formula:
K2O3Te
InChi Key:
BFPJYWDBBLZXOM-UHFFFAOYSA-L
SMILES:
[K+].[K+].[O-][Te]([O-])=O