missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tellurite, MP Biomedicals

Product Code. 11423110 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11423110

Brand: MP Biomedicals 0210042780

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

CAS 7790-58-1
Quantity 100 g
Molecular Formula K2O3Te
Synonym potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e
InChI Key BFPJYWDBBLZXOM-UHFFFAOYSA-L
SMILES [O-][Te](=O)[O-].[K+].[K+]
IUPAC Name dipotassium;tellurite
Molecular Weight (g/mol) 253.794
PubChem CID 65186
ChEBI CHEBI:75248
Formula Weight 253.8
Chemical Name or Material Potassium Tellurite
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium Tellurite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : dipotassium trioxotellurate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.