missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Tellurite MP Biomedicals
Shop All MP Biomedical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| CAS | 7790-58-1 |
| Quantity | 25 g |
| Molecular Formula | K2O3Te |
| Synonym | potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e |
| InChI Key | BFPJYWDBBLZXOM-UHFFFAOYSA-L |
| SMILES | [O-][Te](=O)[O-].[K+].[K+] |
| IUPAC Name | dipotassium;tellurite |
| Molecular Weight (g/mol) | 253.794 |
| PubChem CID | 65186 |
| ChEBI | CHEBI:75248 |
| Show More |
Safety and Handling
Product Identifier
- Potassium Tellurite
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction