accessibility menu, dialog, popup

UserName

CAS RN 77883-43-3

CH3ONNNH2OH3CNNOOOCH3SOHOO

CAS RN 77883-43-3

IUPAC Name: 2-[4-(2,3-dihydro-1,4-benzodioxine-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine; methanesulfonic acid
Synonyms: doxazosin; methanesulfonic acid
Molecular Weight (g/mol): 547.58
Molecular Formula: C24H29N5O8S
InChi Key: VJECBOKJABCYMF-UHFFFAOYNA-N
SMILES: CS(O)(=O)=O.COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1COC2=CC=CC=C2O1
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results