accessibility menu, dialog, popup

UserName

CAS RN 778277-15-9

ONHOHNNNHOFFF

CAS RN 778277-15-9

IUPAC Name: N-(2-hydroxyethyl)-3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Synonyms: N-(2-hydroxyethyl)-3-(6-{[4-(trifluoromethoxy)phenyl]amino}pyrimidin-4-yl)benzamide
Molecular Weight (g/mol): 418.38
Molecular Formula: C20H17F3N4O3
InChi Key: IIQUYGWWHIHOCF-UHFFFAOYSA-N
SMILES: OCCNC(=O)C1=CC=CC(=C1)C1=CC(NC2=CC=C(OC(F)(F)F)C=C2)=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results