missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GNF 5

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective, non-ATP competitive allosteric inhibitor of Bcr-Abl (IC50 = 220 nM for wild-type Abl). Binds the myristate-binding site of Abl. Acts in combination with nilotinib to inhibit T315I Bcr-Abl in vitro and in vivo. Analog of GNF 2 (Cat. No. 4399).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 418.37 |
| Molecular Weight (g/mol) | 418.376 |
| Chemical Name or Material | GNF 5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction