accessibility menu, dialog, popup

UserName

CAS RN 7681-93-8

CH3OOOOHOOHOHOOHOOH3COHNH2OH(E/Z)(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 7681-93-8

IUPAC Name: 22-[(4-amino-3,5-dihydroxy-6-methyloxan-2-yl)oxy]-1,3,26-trihydroxy-12-methyl-10-oxo-6,11,28-trioxatricyclo[22.3.1.0⁵,⁷]octacosa-8,14,16,18,20-pentaene-25-carboxylic acid
Synonyms: natamycin
Molecular Weight (g/mol): 665.73
Molecular Formula: C33H47NO13
InChi Key: NCXMLFZGDNKEPB-UHFFFAOYNA-N
SMILES: CC1OC(OC2CC3OC(O)(CC(O)C3C(O)=O)CC(O)CC3OC3C=CC(=O)OC(C)CC=CC=CC=CC=C2)C(O)C(N)C1O
Color 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results