accessibility menu, dialog, popup

UserName

CAS RN 75998-29-7

CH3OSOClCl

CAS RN 75998-29-7

IUPAC Name: 3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride
Synonyms: 3-chloro-6-methoxy-1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 261.12
Molecular Formula: C10H6Cl2O2S
InChi Key: DTMXTCKEPQWRPO-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(Cl)=C(SC2=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results