CAS RN 741709-57-9
CAS RN 741709-57-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)-2-Pyridinecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.