Learn More
5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)-2-Pyridinecarboxylic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-Yl)-2-Pyridinecarboxylic Acid Ethyl Ester |
| CAS | 741709-57-9 |
| Molecular Formula | C14 H20 B N O4 |
| Quantity | 10 mg |
| Synonym | Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridinecarboxylate, Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate |
| InChI Formula | InChI=1S/C14H20BNO4/c1-6-18-12(17)11-8-7-10(9-16-11)15-19-13(2,3)14(4,5)20-15/h7-9H,6H2,1-5H3 |
| SMILES | CCOC(=O)c1ccc(cn1)B2OC(C)(C)C(C)(C)O2 |
| IUPAC Name | ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxylate |
| Molecular Weight (g/mol) | 277.124 |
| Formula Weight | 277.149 |