CAS RN 73205-13-7
CAS RN 73205-13-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6alpha,9-Difluoro-11beta-hydroxy-16alpha-methyl-17-[(methylsulfanyl)carbonyl]-3-oxoandrosta-1,4-dien-17alpha-yl Propanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.